missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Isocyanate 99.0+%, TCI America™

$96.26 - $281.08

Chemical Identifiers

CAS 3173-56-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00009701
InChI Key YDNLNVZZTACNJX-UHFFFAOYSA-N
Synonym benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate
PubChem CID 76639
IUPAC Name (isocyanatomethyl)benzene
SMILES O=C=NCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I04265G
View Documents
TCI America
I04265G
5 g
Each for $96.26
Only null left
 
I042625G
View Documents
TCI America
I042625G
25 g
Each for $281.08
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3173-56-6
133.15
YDNLNVZZTACNJX-UHFFFAOYSA-N
76639
O=C=NCC1=CC=CC=C1
C8H7NO
MFCD00009701
benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate
(isocyanatomethyl)benzene

Specifications

3173-56-6
118°C
MFCD00009701
2478
YDNLNVZZTACNJX-UHFFFAOYSA-N
(isocyanatomethyl)benzene
76639
≥99.0% (GC)
Benzyl Isocyanate
Yellow
C8H7NO
5 g
benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate
O=C=NCC1=CC=CC=C1
133.15
133.15
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents