missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Isocyanate 99.0+%, TCI America™
Supplier: TCI America I04265G
Specifications
| Benzyl Isocyanate | |
| Yellow | |
| C8H7NO | |
| 5 g | |
| benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate | |
| O=C=NCC1=CC=CC=C1 | |
| 133.15 | |
| 133.15 | |
| Liquid |
| 3173-56-6 | |
| 118°C | |
| MFCD00009701 | |
| 2478 | |
| YDNLNVZZTACNJX-UHFFFAOYSA-N | |
| (isocyanatomethyl)benzene | |
| 76639 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 3173-56-6 | |
| 133.15 | |
| YDNLNVZZTACNJX-UHFFFAOYSA-N | |
| 76639 | |
| O=C=NCC1=CC=CC=C1 |
| C8H7NO | |
| MFCD00009701 | |
| benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate | |
| (isocyanatomethyl)benzene |
Safety and Handling
TSCA : No