missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Dimethylphosphonoacetate 97.0+%, TCI America™

Catalog No. B28155G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28155G 5 g
B28151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28155G Supplier TCI America Supplier No. B28155G

Chemical Identifiers

CAS 57443-18-2
Molecular Formula C11H15O5P
Molecular Weight (g/mol) 258.21
InChI Key QYLGNJMIOVHLQQ-UHFFFAOYSA-N
Synonym benzyl dimethylphosphonoacetate, benzyl 2-dimethoxyphosphoryl acetate, benzyl dimethyl phosphonoacetate, dimethyl benzyloxycarbonyl methyl phosphonate, acetic acid, dimethoxyphosphinyl-, phenylmethyl ester, acmc-209ly9, benzyl dimethylphosphonoethanoate, dimethyl cbz-methyl phosphonate, benzyl dimethoxyphosphoryl acetate, benzyl-2-dimethoxyphosphoryl acetate
PubChem CID 2773759
IUPAC Name benzyl 2-dimethoxyphosphorylacetate
SMILES COP(=O)(CC(=O)OCC1=CC=CC=C1)OC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 258.21
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Dimethylphosphonoacetate