missing translation for 'onlineSavingsMsg'
Learn More

Benzyl dimethyl phosphonoacetate, 98%, Thermo Scientific™

Chemical Identifiers

CAS 57443-18-2
Molecular Formula C11H15O5P
Molecular Weight (g/mol) 258.21
MDL Number MFCD01872507
InChI Key QYLGNJMIOVHLQQ-UHFFFAOYSA-N
Synonym benzyl dimethylphosphonoacetate, benzyl 2-dimethoxyphosphoryl acetate, benzyl dimethyl phosphonoacetate, dimethyl benzyloxycarbonyl methyl phosphonate, acetic acid, dimethoxyphosphinyl-, phenylmethyl ester, acmc-209ly9, benzyl dimethylphosphonoethanoate, dimethyl cbz-methyl phosphonate, benzyl dimethoxyphosphoryl acetate, benzyl-2-dimethoxyphosphoryl acetate
PubChem CID 2773759
IUPAC Name benzyl 2-dimethoxyphosphorylacetate
SMILES COP(=O)(CC(=O)OCC1=CC=CC=C1)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC387370050
SDS View Documents
Thermo Scientific Chemicals
387370050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC387370250
SDS View Documents
Thermo Scientific Chemicals
387370250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for C-C Bond Formation (Olefination)
Specifications

Chemical Identifiers

57443-18-2
258.21
QYLGNJMIOVHLQQ-UHFFFAOYSA-N
2773759
COP(=O)(CC(=O)OCC1=CC=CC=C1)OC
C11H15O5P
MFCD01872507
benzyl dimethylphosphonoacetate, benzyl 2-dimethoxyphosphoryl acetate, benzyl dimethyl phosphonoacetate, dimethyl benzyloxycarbonyl methyl phosphonate, acetic acid, dimethoxyphosphinyl-, phenylmethyl ester, acmc-209ly9, benzyl dimethylphosphonoethanoate, dimethyl cbz-methyl phosphonate, benzyl dimethoxyphosphoryl acetate, benzyl-2-dimethoxyphosphoryl acetate
benzyl 2-dimethoxyphosphorylacetate

Specifications

57443-18-2
100.0
Authentic
Glass bottle
(CH3O)2P(O)CH2C(O)OCH2C5H5
MFCD01872507
benzyl dimethylphosphonoacetate, benzyl 2-dimethoxyphosphoryl acetate, benzyl dimethyl phosphonoacetate, dimethyl benzyloxycarbonyl methyl phosphonate, acetic acid, dimethoxyphosphinyl-, phenylmethyl ester, acmc-209ly9, benzyl dimethylphosphonoethanoate, dimethyl cbz-methyl phosphonate, benzyl dimethoxyphosphoryl acetate, benzyl-2-dimethoxyphosphoryl acetate
COP(=O)(CC(=O)OCC1=CC=CC=C1)OC
258.21
258.21
Benzyl dimethyl phosphonoacetate
97.5
148.0°C to 150.0°C (0.1mmHg)
97.5% min. (HPLC)
C11H15O5P
1.5050 to 1.5070
5g
QYLGNJMIOVHLQQ-UHFFFAOYSA-N
benzyl 2-dimethoxyphosphorylacetate
2773759
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only