missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Chloromethyl Sulfide 96.0+%, TCI America™

Catalog No. b193725g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B193725G 25 g
B19375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B193725G Supplier TCI America Supplier No. B193725G

Chemical Identifiers

CAS 3970-13-6
Molecular Formula C8H9ClS
Molecular Weight (g/mol) 172.67
MDL Number MFCD02093436
InChI Key UMFDSFPDTLREFS-UHFFFAOYSA-N
Synonym alpha-(Chloromethylthio)toluene
PubChem CID 319930
IUPAC Name chloromethylsulfanylmethylbenzene
SMILES C1=CC=C(C=C1)CSCCl

Specifications

Color Yellow
Quantity 25 g
Formula Weight 172.67
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Chloromethyl Sulfide
TSCA No
Recommended Storage Refrigerator