missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloromethyl ether, tech. 70%

Catalog No. AAH60207AD
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60207AD 50 mL
AAH60207AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60207AD Supplier Thermo Scientific Chemicals Supplier No. H60207AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3587-60-8 | C8H9ClO | 156.609 g/mol

Benzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl chloromethyl ether is employed in the introduction of benzoyloxy- and hydroxymethyl groups.

Solubility
Soluble in water [2.7 g/L (25 C)]

Notes
Store at 2-8°C. Keep away from oxidizing agents.

Chemical Identifiers

CAS 3587-60-8
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00000886
InChI Key LADPCMZCENPFGV-UHFFFAOYSA-N
Synonym benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl
PubChem CID 137983
IUPAC Name chloromethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCCl

Specifications

Density 1.126
Boiling Point 102°C
Refractive Index 1.527
Linear Formula C6H5CH2OCH2Cl
Quantity 50 mL
UN Number UN2810
Beilstein 1364034
Solubility Information Soluble in water [2.7g/L (25 C)]
Formula Weight 156.61
Percent Purity 70%
Grade Technical
Chemical Name or Material Benzyl chloromethyl ether
Show More Show Less
Hazard Category H227-H302+H312+H332-H315-H319-H334-H335-H350
Hazard Statement GHS H Statement
H331-H350-H373-H318-H302-H315-H335
Toxic if inhaled.
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye damage.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P235-P261-P264b-P270-P271-P280i-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P332+P313-P342+P311-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only