missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Chloromethyl Ether 90.0+%, TCI America™

Supplier:  TCI America B1207500G

 View more versions of this product

Catalog No. B1207500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Benzyl Chloromethyl Ether
Colorless
C8H9ClO
500 g
benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl
C1=CC=C(C=C1)COCCl
156.609
156.61
Liquid
3587-60-8
56°C
MFCD00000886
2810
LADPCMZCENPFGV-UHFFFAOYSA-N
chloromethoxymethylbenzene
137983
≥90.0% (GC)

Chemical Identifiers

3587-60-8
156.609
LADPCMZCENPFGV-UHFFFAOYSA-N
137983
C1=CC=C(C=C1)COCCl
C8H9ClO
MFCD00000886
benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl
chloromethoxymethylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions