missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Chloromethyl Ether 90.0+%, TCI America™

Catalog No. B120725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B120725G 25 g
B1207100G 100 g
B1207500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B120725G Supplier TCI America Supplier No. B120725G

Chemical Identifiers

CAS 3587-60-8
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00000886
InChI Key LADPCMZCENPFGV-UHFFFAOYSA-N
Synonym benzyl chloromethyl ether, chloromethoxymethyl benzene, chloromethoxy methyl benzene, benzyloxymethyl chloride, chloromethoxymethyl-benzene, benzyl chloromethylether, benzene, chloromethoxy methyl
PubChem CID 137983
IUPAC Name chloromethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCCl

Specifications

Color Colorless
Boiling Point 56°C
Quantity 25 g
UN Number 2810
Formula Weight 156.61
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Chloromethyl Ether
TSCA No
Recommended Storage Refrigerator