missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloroacetate, 98%

Catalog No. p-65786
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC336160050 5 mL Glass bottle
AC336160250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC336160050 Supplier Thermo Scientific Chemicals Supplier No. 336160050

CAS: 140-18-1 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140-18-1
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00157062
InChI Key SOGXBRHOWDEKQB-UHFFFAOYSA-N
Synonym benzyl chloroacetate, benzyl monochloracetate, acetic acid, chloro-, phenylmethyl ester, chloroacetic acid, benzyl ester, monochloroacetate, chloroacetic acid benzyl ester, benzyl-alpha-chloroacetate, phenylmethyl chloroacetate, acetic acid, chloro-, benzyl ester, phenylmethyl 2-chloroacetate
PubChem CID 8786
IUPAC Name benzyl 2-chloroacetate
SMILES C1=CC=C(C=C1)COC(=O)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 1.2200g/mL
Boiling Point 132°C (12.0 mmHg)
Flash Point >110°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OC(O)CH2Cl
Quantity 5 mL
Specific Gravity 1.22
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 184.62
Percent Purity 98%
Chemical Name or Material Benzyl chloroacetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-400-0
RTECSNumber AF8750000
TSCA TSCA

RUO – Research Use Only