missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Carbazate 98.0+%, TCI America™
Supplier: TCI America C15645G
Specifications
| Benzyl Carbazate | |
| 69°C | |
| C8H10N2O2 | |
| 5 g | |
| RXUBZLMIGSAPEJ-UHFFFAOYSA-N | |
| benzyl N-aminocarbamate | |
| 79242 | |
| ≥98.0% (T) |
| 5331-43-1 | |
| White-Yellow | |
| MFCD00041890 | |
| benzyl carbazate, benzyl hydrazinecarboxylate, hydrazinecarboxylic acid, phenylmethyl ester, carbobenzoxyhydrazide, carbazic acid, benzyl ester, benzyloxycarbonyl hydrazide, benzylcarbazate, benzyloxy carbonyl hydrazine, n-cbz-hydrazine, unii-h7gm2h55tc | |
| C1=CC=C(C=C1)COC(=O)NN | |
| 166.18 | |
| 166.18 | |
| Crystalline Powder |
Chemical Identifiers
| 5331-43-1 | |
| 166.18 | |
| RXUBZLMIGSAPEJ-UHFFFAOYSA-N | |
| 79242 | |
| C1=CC=C(C=C1)COC(=O)NN |
| C8H10N2O2 | |
| MFCD00041890 | |
| benzyl carbazate, benzyl hydrazinecarboxylate, hydrazinecarboxylic acid, phenylmethyl ester, carbobenzoxyhydrazide, carbazic acid, benzyl ester, benzyloxycarbonyl hydrazide, benzylcarbazate, benzyloxy carbonyl hydrazine, n-cbz-hydrazine, unii-h7gm2h55tc | |
| benzyl N-aminocarbamate |
Safety and Handling
EINECSNumber : (3)-4377
RTECSNumber : MV1724950
TSCA : Yes