missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Carbazate 98.0+%, TCI America™

Catalog No. p-7125063
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15645G 5 g
C156425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15645G Supplier TCI America Supplier No. C15645G
Only null left

Chemical Identifiers

CAS 5331-43-1
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00041890
InChI Key RXUBZLMIGSAPEJ-UHFFFAOYSA-N
Synonym benzyl carbazate, benzyl hydrazinecarboxylate, hydrazinecarboxylic acid, phenylmethyl ester, carbobenzoxyhydrazide, carbazic acid, benzyl ester, benzyloxycarbonyl hydrazide, benzylcarbazate, benzyloxy carbonyl hydrazine, n-cbz-hydrazine, unii-h7gm2h55tc
PubChem CID 79242
IUPAC Name benzyl N-aminocarbamate
SMILES C1=CC=C(C=C1)COC(=O)NN

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl Carbazate
EINECSNumber (3)-4377
RTECSNumber MV1724950
TSCA Yes