missing translation for 'onlineSavingsMsg'
Learn More

Benzyl carbamate, 99%

CAS: 621-84-1 | C8H9NO2 | 151.17 g/mol

$78.86 - $433.49

Chemical Identifiers

CAS 621-84-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007965
InChI Key PUJDIJCNWFYVJX-UHFFFAOYSA-N
Synonym carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421
PubChem CID 12136
IUPAC Name benzyl carbamate
SMILES NC(=O)OCC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1156914
View Documents Promotion Details
N/A 25 g
Each for $78.86
Only null left
 
AAA1156922
View Documents Promotion Details
N/A 100 g
Each for $212.40
Only null left
 
AAA1156930
View Documents Promotion Details
N/A 250 g
Each for $433.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

621-84-1
151.17
PUJDIJCNWFYVJX-UHFFFAOYSA-N
12136
NC(=O)OCC1=CC=CC=C1
C8H9NO2
MFCD00007965
carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421
benzyl carbamate

Specifications

85°C to 89°C
C8H9NO2
1865635
PUJDIJCNWFYVJX-UHFFFAOYSA-N
benzyl carbamate
12136
99%
621-84-1
MFCD00007965
carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421
NC(=O)OCC1=CC=CC=C1
151.17
151.17
Benzyl carbamate
Videos
Safety and Handling

Safety and Handling

P261-P271-P304+P340-P312-P501c

H332-H335

EINECSNumber : 210-710-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only