missing translation for 'onlineSavingsMsg'
Learn More

Benzyl carbamate, 98%

Catalog No. AC105890250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105890250 25 g Glass Bottle
AC105891000 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105890250 Supplier Thermo Scientific Chemicals Supplier No. 105890250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 621-84-1 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 621-84-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007965
InChI Key PUJDIJCNWFYVJX-UHFFFAOYSA-N
Synonym carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421
PubChem CID 12136
IUPAC Name benzyl carbamate
SMILES NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 86.0°C to 89.0°C
CAS Min % 97.5
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula H2NCO2CH2C6H5
Beilstein 06, 437
Fieser 13,79
Quantity 25 g
Formula Weight 151.16
Percent Purity 98%
Physical Form Powder or Flakes
Chemical Name or Material Benzyl carbamate, 99%
Show More Show Less
EINECSNumber 210-710-4

RUO – Research Use Only