missing translation for 'onlineSavingsMsg'
Learn More

Benzyl carbamate, 98%

CAS: 621-84-1 | C8H9NO2 | 151.17 g/mol

$77.93

Chemical Identifiers

CAS 621-84-1
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007965
InChI Key PUJDIJCNWFYVJX-UHFFFAOYSA-N
Synonym carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421
PubChem CID 12136
IUPAC Name benzyl carbamate
SMILES NC(=O)OCC1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC105890250
View Documents Promotion Details
N/A 25 g Glass Bottle
Each for $77.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

621-84-1
151.17
PUJDIJCNWFYVJX-UHFFFAOYSA-N
12136
NC(=O)OCC1=CC=CC=C1
C8H9NO2
MFCD00007965
carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421
benzyl carbamate

Specifications

86.0°C to 89.0°C
97.5
White to Beige
97.5% min. (GC)
H2NCO2CH2C6H5
06, 437
carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421
NC(=O)OCC1=CC=CC=C1
151.17
151.16
Powder or Flakes
621-84-1
100.0
Authentic
C8H9NO2
MFCD00007965
13,79
PUJDIJCNWFYVJX-UHFFFAOYSA-N
benzyl carbamate
12136
98%
Benzyl carbamate, 99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-710-4

SDS
Documents

Documents

RUO – Research Use Only