missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Butyl Phthalate 97.0+%, TCI America™

Catalog No. p-7134535
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P028825G 25 g
P0288500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P028825G Supplier TCI America Supplier No. P028825G
Only null left

Chemical Identifiers

CAS 85-68-7
Molecular Formula C19H20O4
Molecular Weight (g/mol) 312.365
MDL Number MFCD00009440
InChI Key IRIAEXORFWYRCZ-UHFFFAOYSA-N
Synonym Phthalic Acid Benzyl Butyl Ester, BBP
PubChem CID 2347
ChEBI CHEBI:34595
IUPAC Name 2-O-benzyl 1-O-butyl benzene-1,2-dicarboxylate
SMILES CCCCOC(=O)C1=CC=CC=C1C(=O)OCC2=CC=CC=C2

Specifications

Physical Form Liquid
UN Number 3082
Color Yellow
Boiling Point 250°C
Quantity 25 g
Formula Weight 312.37
Percent Purity ≥97.0% (GC)
Chemical Name or Material Benzyl Butyl Phthalate
EINECSNumber (3)-1312
RTECSNumber TH9990000
TSCA Yes