missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Butyl Phthalate 97.0+%, TCI America™
Specifications
Specifications
| Physical Form | Liquid |
| UN Number | 3082 |
| Color | Yellow |
| Boiling Point | 250°C |
| Quantity | 25 g |
| Formula Weight | 312.37 |
| Percent Purity | ≥97.0% (GC) |
| Chemical Name or Material | Benzyl Butyl Phthalate |
Safety and Handling
| EINECSNumber | (3)-1312 |
| RTECSNumber | TH9990000 |
| TSCA | Yes |