missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Butyl Ether 95.0+%, TCI America™
$85.75 - $279.60
Chemical Identifiers
| CAS | 588-67-0 |
|---|---|
| Molecular Formula | C11H16O |
| Molecular Weight (g/mol) | 164.25 |
| MDL Number | MFCD00039954 |
| InChI Key | MAYUYFCAPVDYBQ-UHFFFAOYSA-N |
| Synonym | benzyl butyl ether, ether, benzyl butyl, butyl benzyl ether, benzyl n-butyl ether, benzene, butoxymethyl, butoxymethyl benzene, n-butyl benzyl ether, unii-8483x8zo8n, ether, benzyl n-butyl, butoxyphenylmethane |
| PubChem CID | 61134 |
| IUPAC Name | (butoxymethyl)benzene |
| SMILES | CCCCOCC1=CC=CC=C1 |
Chemical Identifiers
| 588-67-0 | |
| 164.25 | |
| MAYUYFCAPVDYBQ-UHFFFAOYSA-N | |
| 61134 | |
| CCCCOCC1=CC=CC=C1 |
| C11H16O | |
| MFCD00039954 | |
| benzyl butyl ether, ether, benzyl butyl, butyl benzyl ether, benzyl n-butyl ether, benzene, butoxymethyl, butoxymethyl benzene, n-butyl benzyl ether, unii-8483x8zo8n, ether, benzyl n-butyl, butoxyphenylmethane | |
| (butoxymethyl)benzene |
Specifications
| 588-67-0 | |
| 220°C | |
| MFCD00039954 | |
| benzyl butyl ether, ether, benzyl butyl, butyl benzyl ether, benzyl n-butyl ether, benzene, butoxymethyl, butoxymethyl benzene, n-butyl benzyl ether, unii-8483x8zo8n, ether, benzyl n-butyl, butoxyphenylmethane | |
| CCCCOCC1=CC=CC=C1 | |
| 164.25 | |
| 164.25 | |
| Liquid |
| Yellow | |
| C11H16O | |
| 5 g | |
| MAYUYFCAPVDYBQ-UHFFFAOYSA-N | |
| (butoxymethyl)benzene | |
| 61134 | |
| ≥95.0% (GC) | |
| Benzyl Butyl Ether |
Safety and Handling
TSCA : No