missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Bromoacetate 96.0+%, TCI America™
$41.02 - $361.62
Chemical Identifiers
| CAS | 5437-45-6 |
|---|---|
| Molecular Formula | C9H9BrO2 |
| Molecular Weight (g/mol) | 229.073 |
| MDL Number | MFCD00000190 |
| InChI Key | JHVLLYQQQYIWKX-UHFFFAOYSA-N |
| Synonym | benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate |
| PubChem CID | 62576 |
| IUPAC Name | benzyl 2-bromoacetate |
| SMILES | C1=CC=C(C=C1)COC(=O)CBr |
Chemical Identifiers
| 5437-45-6 | |
| 229.073 | |
| JHVLLYQQQYIWKX-UHFFFAOYSA-N | |
| 62576 | |
| C1=CC=C(C=C1)COC(=O)CBr |
| C9H9BrO2 | |
| MFCD00000190 | |
| benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate | |
| benzyl 2-bromoacetate |
Specifications
| 5437-45-6 | |
| 170°C | |
| MFCD00000190 | |
| benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate | |
| C1=CC=C(C=C1)COC(=O)CBr | |
| 229.073 | |
| 229.07 | |
| Liquid |
| Yellow | |
| C9H9BrO2 | |
| 25 g | |
| JHVLLYQQQYIWKX-UHFFFAOYSA-N | |
| benzyl 2-bromoacetate | |
| 62576 | |
| ≥96.0% (GC) | |
| Benzyl Bromoacetate |
Safety and Handling
EINECSNumber : (9)-1875
RTECSNumber : AF5957215
TSCA : No