Learn More
Benzyl bromoacetate, 97%
CAS: 5437-45-6 | C9H9BrO2 | 229.073 g/mol
$70.78 - $637.27
Chemical Identifiers
| CAS | 5437-45-6 |
|---|---|
| Molecular Formula | C9H9BrO2 |
| Molecular Weight (g/mol) | 229.073 |
| MDL Number | MFCD00000190 |
| InChI Key | JHVLLYQQQYIWKX-UHFFFAOYSA-N |
| Synonym | benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate |
| PubChem CID | 62576 |
| IUPAC Name | benzyl 2-bromoacetate |
| SMILES | C1=CC=C(C=C1)COC(=O)CBr |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1763018
|
Thermo Scientific Chemicals
A1763018 |
50 g |
Each for $70.78
|
|
|||||
|
AAA1763030
|
Thermo Scientific Chemicals
A1763030 |
250 g |
Each for $225.96
|
|
|||||
|
AAA176300B
|
Thermo Scientific Chemicals
A176300B |
1000 g |
Each for $637.27
|
|
|||||
Description
It is employed in biological research purpose. Benzyl bromoacetate was used in the alkylation of (-)-2,3-O-isopropylidene-D-threitol that afforded lipopeptide, 2-[(4R,5R)-5-({[(9H-fluoren-9-yl)methoxy]carbonylaminomethyl}-2,2-dimethyl-1,3-dioxolan-4-yl)methoxy]acetic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed in biological research purpose. Benzyl bromoacetate was used in the alkylation of (-)-2,3-O-isopropylidene-D-threitol that afforded lipopeptide, 2-[(4R,5R)-5-({[(9H-fluoren-9-yl)methoxy]carbonylaminomethyl}-2,2-dimethyl-1,3-dioxolan-4-yl)methoxy]acetic acid.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place. Store under 20ËC Freezer, Under Inert Atmosphere.
Chemical Identifiers
| 5437-45-6 | |
| 229.073 | |
| JHVLLYQQQYIWKX-UHFFFAOYSA-N | |
| 62576 | |
| C1=CC=C(C=C1)COC(=O)CBr |
| C9H9BrO2 | |
| MFCD00000190 | |
| benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate | |
| benzyl 2-bromoacetate |
Specifications
| 5437-45-6 | |
| 157°C to 159°C (10 mmHg) | |
| Pungent | |
| 1.544 | |
| 50 g | |
| benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate | |
| JHVLLYQQQYIWKX-UHFFFAOYSA-N | |
| benzyl 2-bromoacetate | |
| 62576 | |
| 97% |
| 1.46 | |
| >110°C (230°F) | |
| C9H9BrO2 | |
| MFCD00000190 | |
| 973658 | |
| Not miscible or difficult to mix in water. | |
| C1=CC=C(C=C1)COC(=O)CBr | |
| 229.073 | |
| 229.08 | |
| Benzyl bromoacetate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 226-611-4
RTECSNumber : AF5957215
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only