missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Bromide (stabilized with Propylene Oxide) 98.0+%, TCI America™

Catalog No. B041125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B041125G 25 g
B0411100G 100 g
B0411500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B041125G Supplier TCI America Supplier No. B041125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100-39-0
Molecular Formula C7H7Br
Molecular Weight (g/mol) 171.037
MDL Number MFCD00000172
InChI Key AGEZXYOZHKGVCM-UHFFFAOYSA-N
Synonym benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
PubChem CID 7498
ChEBI CHEBI:59858
IUPAC Name bromomethylbenzene
SMILES C1=CC=C(C=C1)CBr

Specifications

Color Yellow
Boiling Point 199°C
Quantity 25 g
UN Number 1737
Formula Weight 171.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Bromide (stabilized with Propylene Oxide)
EINECSNumber (3)-2689
RTECSNumber XS7965000
TSCA Yes