missing translation for 'onlineSavingsMsg'
Learn More

Benzyl bromide, 99%

CAS: 100-39-0 | C7H7Br | 171.037 g/mol

Supplier:  Thermo Scientific Chemicals A135350B

 View more versions of this product

Catalog No. AAA135350B


Only null left
Explore available promotions

Description

Description

Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographic chemicals, synthetic resins, dyes and plasticizers and esters. It is also involved in the synthesis of (E)-(2-Iodovinyl)benzene by reacting it with diiodomethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographi
Specifications

Specifications

Benzyl bromide
-4°C
198°C to 199°C
Pungent, Makes Eyes Water
1.575
1000 g
385801
14,1128
Miscible with benzene,carbon tetrachloride,ethanol and ether.
C1=CC=C(C=C1)CBr
171.037
CHEBI:59858
99%
100-39-0
1.438
87°C (189°F)
C7H7Br
MFCD00000172
UN1737
Moisture sensitive
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
AGEZXYOZHKGVCM-UHFFFAOYSA-N
bromomethylbenzene
7498
171.04
Liquid

Chemical Identifiers

100-39-0
171.037
AGEZXYOZHKGVCM-UHFFFAOYSA-N
7498
bromomethylbenzene
C7H7Br
MFCD00000172
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
CHEBI:59858
C1=CC=C(C=C1)CBr
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H335-H227-H319
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes serious eye irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BENZYL BROMIDE

EINECSNumber : 202-847-3

RTECSNumber : XS7965000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only