missing translation for 'onlineSavingsMsg'
Learn More

Medisca Benzyl Alcohol NF

Catalog No. 14185493
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14185493 500 mL
14185489 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14185493 Supplier Medisca Supplier No. 102908

Benzyl Alcohol NF is a clear, colorless, oily liquid. It is hygroscopic.

Benzyl Alcohol NF is a clear, colorless, oily liquid. It is hygroscopic.

Alternative name(s): Benzenemethanol, Benzene methanol, Phenylcarbinol, Phenylmethanol, alpha-Hydroxytoluene

Specifications

Chemical Name or Material Benzyl Alcohol NF
Packaging Bottle, Amber Glass, Boston Round, 500mL, 38-439
CAS 100-51-6
Type Drug Standard
Molecular Weight (g/mol) 108.14
Quantity 500 mL
Percent Purity 98 to 100.5 %
Grade NF