missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medisca Benzyl Alcohol NF
Description
Benzyl Alcohol NF is a clear, colorless, oily liquid. It is hygroscopic.
Alternative name(s): Benzenemethanol, Benzene methanol, Phenylcarbinol, Phenylmethanol, alpha-Hydroxytoluene
Specifications
Specifications
| Chemical Name or Material | Benzyl Alcohol NF |
| Packaging | Jug, 4L, Amber Glass, Round, 38-439 (for MCP38439LNC) |
| CAS | 100-51-6 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 108.14 |
| Quantity | 4 L |
| Percent Purity | 98 to 100.5 % |
| Grade | NF |