missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Acrylate (stabilized with MEHQ) 97.0+%, TCI America™
Supplier: TCI America B462725G
Specifications
| Benzyl Acrylate (stabilized with MEHQ) | |
| Colorless | |
| C10H10O2 | |
| 25 g | |
| GCTPMLUUWLLESL-UHFFFAOYSA-N | |
| benzyl prop-2-enoate | |
| 75617 | |
| ≥97.0% (GC) |
| 2495-35-4 | |
| 60°C | |
| MFCD00048147 | |
| benzyl acrylate, benzylacrylate, 2-propenoic acid, phenylmethyl ester, sartomer sr 432, acrylic acid, benzyl ester, acrylic acid benzyl ester, phenylmethyl prop-2-enoate, benzyl 2-propenoate, fancryl fa-bza, acrylic acid benzyl | |
| C=CC(=O)OCC1=CC=CC=C1 | |
| 162.19 | |
| 162.19 | |
| Liquid |
Chemical Identifiers
| 2495-35-4 | |
| 162.19 | |
| GCTPMLUUWLLESL-UHFFFAOYSA-N | |
| 75617 | |
| C=CC(=O)OCC1=CC=CC=C1 |
| C10H10O2 | |
| MFCD00048147 | |
| benzyl acrylate, benzylacrylate, 2-propenoic acid, phenylmethyl ester, sartomer sr 432, acrylic acid, benzyl ester, acrylic acid benzyl ester, phenylmethyl prop-2-enoate, benzyl 2-propenoate, fancryl fa-bza, acrylic acid benzyl | |
| benzyl prop-2-enoate |
Safety and Handling
EINECSNumber : (3)-2873
TSCA : Yes