missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Acrylate (stabilized with MEHQ) 97.0+%, TCI America™

Catalog No. p-7123653
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B462725G 25 g
B4627100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B462725G Supplier TCI America Supplier No. B462725G
Only null left

Chemical Identifiers

CAS 2495-35-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00048147
InChI Key GCTPMLUUWLLESL-UHFFFAOYSA-N
Synonym benzyl acrylate, benzylacrylate, 2-propenoic acid, phenylmethyl ester, sartomer sr 432, acrylic acid, benzyl ester, acrylic acid benzyl ester, phenylmethyl prop-2-enoate, benzyl 2-propenoate, fancryl fa-bza, acrylic acid benzyl
PubChem CID 75617
IUPAC Name benzyl prop-2-enoate
SMILES C=CC(=O)OCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 60°C
Quantity 25 g
Formula Weight 162.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Acrylate (stabilized with MEHQ)
EINECSNumber (3)-2873
TSCA Yes