missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Acetate 99.0+%, TCI America™
$39.14 - $63.38
Chemical Identifiers
| CAS | 140-11-4 |
|---|---|
| Molecular Formula | C9H10O2 |
| Molecular Weight (g/mol) | 150.177 |
| MDL Number | MFCD00008712 |
| InChI Key | QUKGYYKBILRGFE-UHFFFAOYSA-N |
| Synonym | acetic acid benzyl ester, benzyl ethanoate, acetic acid, phenylmethyl ester, phenylmethyl acetate, acetic acid, benzyl ester, alpha-acetoxytoluene, phenylmethyl ethanoate, acetoxymethyl benzene, benzylester kyseliny octove, caswell no. 081ea |
| PubChem CID | 8785 |
| ChEBI | CHEBI:52051 |
| IUPAC Name | benzyl acetate |
| SMILES | CC(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 140-11-4 | |
| 150.177 | |
| QUKGYYKBILRGFE-UHFFFAOYSA-N | |
| 8785 | |
| benzyl acetate |
| C9H10O2 | |
| MFCD00008712 | |
| acetic acid benzyl ester, benzyl ethanoate, acetic acid, phenylmethyl ester, phenylmethyl acetate, acetic acid, benzyl ester, alpha-acetoxytoluene, phenylmethyl ethanoate, acetoxymethyl benzene, benzylester kyseliny octove, caswell no. 081ea | |
| CHEBI:52051 | |
| CC(=O)OCC1=CC=CC=C1 |
Specifications
| 140-11-4 | |
| Yellow | |
| C9H10O2 | |
| 25 g | |
| QUKGYYKBILRGFE-UHFFFAOYSA-N | |
| benzyl acetate | |
| 8785 | |
| 150.18 | |
| Liquid |
| -51°C | |
| 214°C | |
| MFCD00008712 | |
| acetic acid benzyl ester, benzyl ethanoate, acetic acid, phenylmethyl ester, phenylmethyl acetate, acetic acid, benzyl ester, alpha-acetoxytoluene, phenylmethyl ethanoate, acetoxymethyl benzene, benzylester kyseliny octove, caswell no. 081ea | |
| CC(=O)OCC1=CC=CC=C1 | |
| 150.177 | |
| CHEBI:52051 | |
| ≥99.0% (GC) | |
| Benzyl Acetate |
Safety and Handling
EINECSNumber : (3)-1020&(3)-1045
RTECSNumber : AF5075000
TSCA : Yes