missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Acetate 99.0+%, TCI America™
Supplier: TCI America A0022500G
Specifications
| Benzyl Acetate | |
| -51°C | |
| 214°C | |
| MFCD00008712 | |
| acetic acid benzyl ester, benzyl ethanoate, acetic acid, phenylmethyl ester, phenylmethyl acetate, acetic acid, benzyl ester, alpha-acetoxytoluene, phenylmethyl ethanoate, acetoxymethyl benzene, benzylester kyseliny octove, caswell no. 081ea | |
| CC(=O)OCC1=CC=CC=C1 | |
| 150.177 | |
| CHEBI:52051 | |
| ≥99.0% (GC) |
| 140-11-4 | |
| Yellow | |
| C9H10O2 | |
| 500 g | |
| QUKGYYKBILRGFE-UHFFFAOYSA-N | |
| benzyl acetate | |
| 8785 | |
| 150.18 | |
| Liquid |
Chemical Identifiers
| 140-11-4 | |
| 150.177 | |
| QUKGYYKBILRGFE-UHFFFAOYSA-N | |
| 8785 | |
| benzyl acetate |
| C9H10O2 | |
| MFCD00008712 | |
| acetic acid benzyl ester, benzyl ethanoate, acetic acid, phenylmethyl ester, phenylmethyl acetate, acetic acid, benzyl ester, alpha-acetoxytoluene, phenylmethyl ethanoate, acetoxymethyl benzene, benzylester kyseliny octove, caswell no. 081ea | |
| CHEBI:52051 | |
| CC(=O)OCC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (3)-1020&(3)-1045
RTECSNumber : AF5075000
TSCA : Yes