missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 6-Bromohexyl Ether 97.0+%, TCI America™

Catalog No. p-7119782
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28205G 5 g
B282025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28205G Supplier TCI America Supplier No. B28205G

Chemical Identifiers

CAS 54247-27-7
Molecular Formula C13H19BrO
Molecular Weight (g/mol) 271.20
MDL Number MFCD02258455
InChI Key UHDZRWPKYMHVNV-UHFFFAOYSA-N
Synonym Benzyloxyhexyl Bromide, (6-Bromohexyloxymethyl)benzene
PubChem CID 2784764
IUPAC Name {[(6-bromohexyl)oxy]methyl}benzene
SMILES BrCCCCCCOCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 271.20
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl 6-Bromohexyl Ether