missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-Hydroxyphenyl Ketone 98.0+%, TCI America™

Catalog No. p-7123705
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B47221G 1 g
B47225G 5 g
2 options

Catalog No. B47221G

Supplier: TCI America B47221G

Only null left

Chemical Identifiers

CAS 2491-32-9
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.248
MDL Number MFCD00002360
InChI Key JBQTZLNCDIFCCO-UHFFFAOYSA-N
Synonym benzyl 4-hydroxyphenyl ketone, 1-4-hydroxyphenyl-2-phenylethanone, 4-hydroxydeoxybenzoin, acetophenone, 4'-hydroxy-2-phenyl, 1-4-hydroxyphenyl-2-phenylethan-1-one, ethanone, 1-4-hydroxyphenyl-2-phenyl, acmc-20alpm, 4-hydroxydesoxybenzoin, p-phenylacetyl phenol, maybridge3_000426
PubChem CID 75607
IUPAC Name 1-(4-hydroxyphenyl)-2-phenylethanone
SMILES C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)O

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 1 g
Formula Weight 212.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl 4-Hydroxyphenyl Ketone