accessibility menu, dialog, popup

UserName

OHO

CAS RN 2491-32-9

IUPAC Name: 1-(4-hydroxyphenyl)-2-phenylethan-1-one
Synonyms: 1-(4-hydroxyphenyl)-2-phenylethanone
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: JBQTZLNCDIFCCO-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)