missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-Hydroxybenzoate 98.0+%, TCI America™

Supplier:  TCI America H020925G

 View more versions of this product

Catalog No. H020925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Benzyl 4-Hydroxybenzoate
111°C
C14H12O3
25 g
benzylparaben, benzyl p-hydroxybenzoate, nipabenzyl, parosept, benzyl paraben, benzyl parasept, benzyl tegosept, solbrol z, benzyl-parasept, p-hydroxybenzoic acid benzyl ester
OC1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1
228.25
CHEBI:34571
≥98.0% (GC,T)
94-18-8
White
MFCD00016471
3077
MOZDKDIOPSPTBH-UHFFFAOYSA-N
benzyl 4-hydroxybenzoate
7180
228.25
Crystalline Powder

Chemical Identifiers

94-18-8
228.25
MOZDKDIOPSPTBH-UHFFFAOYSA-N
7180
benzyl 4-hydroxybenzoate
C14H12O3
MFCD00016471
benzylparaben, benzyl p-hydroxybenzoate, nipabenzyl, parosept, benzyl paraben, benzyl parasept, benzyl tegosept, solbrol z, benzyl-parasept, p-hydroxybenzoic acid benzyl ester
CHEBI:34571
OC1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1325

RTECSNumber : DH2691000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions