missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 4-Hydroxybenzoate 98.0+%, TCI America™
Supplier: TCI America H0209100G
Specifications
| Benzyl 4-Hydroxybenzoate | |
| 111°C | |
| C14H12O3 | |
| 100 g | |
| benzylparaben, benzyl p-hydroxybenzoate, nipabenzyl, parosept, benzyl paraben, benzyl parasept, benzyl tegosept, solbrol z, benzyl-parasept, p-hydroxybenzoic acid benzyl ester | |
| OC1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1 | |
| 228.25 | |
| CHEBI:34571 | |
| ≥98.0% (GC,T) |
| 94-18-8 | |
| White | |
| MFCD00016471 | |
| 3077 | |
| MOZDKDIOPSPTBH-UHFFFAOYSA-N | |
| benzyl 4-hydroxybenzoate | |
| 7180 | |
| 228.25 | |
| Crystalline Powder |
Chemical Identifiers
| 94-18-8 | |
| 228.25 | |
| MOZDKDIOPSPTBH-UHFFFAOYSA-N | |
| 7180 | |
| benzyl 4-hydroxybenzoate |
| C14H12O3 | |
| MFCD00016471 | |
| benzylparaben, benzyl p-hydroxybenzoate, nipabenzyl, parosept, benzyl paraben, benzyl parasept, benzyl tegosept, solbrol z, benzyl-parasept, p-hydroxybenzoic acid benzyl ester | |
| CHEBI:34571 | |
| OC1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (9)-1325
RTECSNumber : DH2691000
TSCA : Yes