missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-hydroxybenzoate, 99%

Catalog No. AAA1173509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1173509 10 g
AAA1173518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1173509 Supplier Thermo Scientific Chemicals Supplier No. A1173509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 94-18-8 | C14H12O3 | 228.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94-18-8
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00016471
InChI Key MOZDKDIOPSPTBH-UHFFFAOYSA-N
Synonym benzylparaben, benzyl p-hydroxybenzoate, nipabenzyl, parosept, benzyl paraben, benzyl parasept, benzyl tegosept, solbrol z, benzyl-parasept, p-hydroxybenzoic acid benzyl ester
PubChem CID 7180
ChEBI CHEBI:34571
IUPAC Name benzyl 4-hydroxybenzoate
SMILES OC1=CC=C(C=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 110°C to 114°C
Quantity 10 g
Beilstein 2115995
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material Benzyl 4-hydroxybenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-311-9
RTECSNumber DH2691000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only