missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-Chlorophenyl Ketone 98.0+%, TCI America™

Catalog No. p-7123697
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47045G 5 g
B470425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47045G Supplier TCI America Supplier No. B47045G
Only null left

Chemical Identifiers

CAS 1889-71-0
Molecular Formula C14H11ClO
Molecular Weight (g/mol) 230.69
MDL Number MFCD00016342
InChI Key DXVALSKCLLBZEB-UHFFFAOYSA-N
Synonym benzyl 4-chlorophenyl ketone, 1-4-chlorophenyl-2-phenylethanone, 4'-chloro-2-phenylacetophenone, 4-chlorodeoxybenzoin, 1-4-chlorophenyl-2-phenylethan-1-one, 1-4-chlorophenyl-2-phenyl-ethanone, 4-chlorodesoxy benzoin, chlorodeoxybenzoin, deoxy-4-chlorobenzoin, acmc-20a6by
PubChem CID 233840
IUPAC Name 1-(4-chlorophenyl)-2-phenylethan-1-one
SMILES ClC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1

Specifications

Melting Point 106°C
Color White-Yellow
Boiling Point 187°C
Quantity 5 g
Formula Weight 230.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl 4-Chlorophenyl Ketone