missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-chlorophenyl ketone, 98%

Catalog No. AAA1595906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1595906 5 g
AAA1595914 25 g
AAA1595922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1595906 Supplier Thermo Scientific Chemicals Supplier No. A1595906
Only null left

CAS: 1889-71-0 | C14H11ClO | 230.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1889-71-0
Molecular Formula C14H11ClO
Molecular Weight (g/mol) 230.69
MDL Number MFCD00016342
InChI Key DXVALSKCLLBZEB-UHFFFAOYSA-N
Synonym benzyl 4-chlorophenyl ketone, 1-4-chlorophenyl-2-phenylethanone, 4'-chloro-2-phenylacetophenone, 4-chlorodeoxybenzoin, 1-4-chlorophenyl-2-phenylethan-1-one, 1-4-chlorophenyl-2-phenyl-ethanone, 4-chlorodesoxy benzoin, chlorodeoxybenzoin, deoxy-4-chlorobenzoin, acmc-20a6by
PubChem CID 233840
IUPAC Name 1-(4-chlorophenyl)-2-phenylethanone
SMILES ClC1=CC=C(C=C1)C(=O)CC1=CC=CC=C1

Specifications

Melting Point 103°C to 107°C
Quantity 5 g
Beilstein 976443
Formula Weight 230.69
Percent Purity 98%
Chemical Name or Material Benzyl 4-chlorophenyl ketone
Hazard Category H317-H318
Precautionary Statement P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only