missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-Hydroxyphenyl Ketone 98.0+%, TCI America™

$111.84 - $385.86

Chemical Identifiers

CAS 2491-31-8
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00218589
InChI Key VGHVJQXWDXRTRJ-UHFFFAOYSA-N
Synonym 2′C-Hydroxy-2-phenylacetophenone
PubChem CID 137612
IUPAC Name 1-(2-hydroxyphenyl)-2-phenylethan-1-one
SMILES OC1=CC=CC=C1C(=O)CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B4673200MG
View Documents
TCI America
B4673200MG
200 mg
Each for $111.84
Only null left
 
B46731G
View Documents
TCI America
B46731G
1 g
Each for $385.86
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2491-31-8
212.25
VGHVJQXWDXRTRJ-UHFFFAOYSA-N
137612
OC1=CC=CC=C1C(=O)CC1=CC=CC=C1
C14H12O2
MFCD00218589
2′C-Hydroxy-2-phenylacetophenone
1-(2-hydroxyphenyl)-2-phenylethan-1-one

Specifications

2491-31-8
165°C
MFCD00218589
2′C-Hydroxy-2-phenylacetophenone
OC1=CC=CC=C1C(=O)CC1=CC=CC=C1
212.25
212.25
Crystalline Powder
57°C
C14H12O2
200 mg
VGHVJQXWDXRTRJ-UHFFFAOYSA-N
1-(2-hydroxyphenyl)-2-phenylethan-1-one
137612
≥98.0% (GC)
Benzyl 2-Hydroxyphenyl Ketone
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'