missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 2-Hydroxyphenyl Ketone 98.0+%, TCI America™
Supplier: TCI America B46731G
Specifications
| Benzyl 2-Hydroxyphenyl Ketone | |
| 57°C | |
| C14H12O2 | |
| 1 g | |
| VGHVJQXWDXRTRJ-UHFFFAOYSA-N | |
| 1-(2-hydroxyphenyl)-2-phenylethan-1-one | |
| 137612 | |
| ≥98.0% (GC) |
| 2491-31-8 | |
| 165°C | |
| MFCD00218589 | |
| 2′C-Hydroxy-2-phenylacetophenone | |
| OC1=CC=CC=C1C(=O)CC1=CC=CC=C1 | |
| 212.25 | |
| 212.25 | |
| Crystalline Powder |
Chemical Identifiers
| 2491-31-8 | |
| 212.25 | |
| VGHVJQXWDXRTRJ-UHFFFAOYSA-N | |
| 137612 | |
| OC1=CC=CC=C1C(=O)CC1=CC=CC=C1 |
| C14H12O2 | |
| MFCD00218589 | |
| 2′C-Hydroxy-2-phenylacetophenone | |
| 1-(2-hydroxyphenyl)-2-phenylethan-1-one |
Safety and Handling
TSCA : No