missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 2-Chloroethyl Ether 98.0+%, TCI America™
$227.16 - $671.89
Chemical Identifiers
| CAS | 17229-17-3 |
|---|---|
| Molecular Formula | C9H11ClO |
| Molecular Weight (g/mol) | 170.636 |
| InChI Key | UPIMXDQREQJWMR-UHFFFAOYSA-N |
| Synonym | 2-(Benzyloxy)ethyl Chloride, (2-Chloroethoxymethyl)benzene |
| PubChem CID | 223538 |
| IUPAC Name | 2-chloroethoxymethylbenzene |
| SMILES | C1=CC=C(C=C1)COCCCl |
Chemical Identifiers
| 17229-17-3 | |
| 170.636 | |
| 2-(Benzyloxy)ethyl Chloride, (2-Chloroethoxymethyl)benzene | |
| 2-chloroethoxymethylbenzene |
| C9H11ClO | |
| UPIMXDQREQJWMR-UHFFFAOYSA-N | |
| 223538 | |
| C1=CC=C(C=C1)COCCCl |
Specifications
| 17229-17-3 | |
| C9H11ClO | |
| 2-(Benzyloxy)ethyl Chloride, (2-Chloroethoxymethyl)benzene | |
| C1=CC=C(C=C1)COCCCl | |
| 170.636 | |
| 170.64 | |
| Liquid |
| Yellow | |
| 5 g | |
| UPIMXDQREQJWMR-UHFFFAOYSA-N | |
| 2-chloroethoxymethylbenzene | |
| 223538 | |
| ≥98.0% (GC) | |
| Benzyl 2-Chloroethyl Ether |
Safety and Handling
TSCA : No