missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 2-Bromoethyl Ether 97.0+%, TCI America™
$63.38 - $249.78
Chemical Identifiers
| CAS | 1462-37-9 |
|---|---|
| Molecular Formula | C9H11BrO |
| Molecular Weight (g/mol) | 215.09 |
| MDL Number | MFCD01321307 |
| InChI Key | FWOHDAGPWDEWIB-UHFFFAOYSA-N |
| Synonym | benzyl 2-bromoethyl ether, 2-bromoethoxy methyl benzene, 2-benzyloxy-1-bromoethane, benzyloxyethyl bromide, benzene, 2-bromoethoxy methyl, 1-bromo-2-benzyloxyethane, 2-bromoethoxymethyl benzene, 2-bromo-ethoxymethyl-benzene |
| PubChem CID | 73833 |
| IUPAC Name | 2-bromoethoxymethylbenzene |
| SMILES | C1=CC=C(C=C1)COCCBr |
Chemical Identifiers
| 1462-37-9 | |
| 215.09 | |
| FWOHDAGPWDEWIB-UHFFFAOYSA-N | |
| 73833 | |
| C1=CC=C(C=C1)COCCBr |
| C9H11BrO | |
| MFCD01321307 | |
| benzyl 2-bromoethyl ether, 2-bromoethoxy methyl benzene, 2-benzyloxy-1-bromoethane, benzyloxyethyl bromide, benzene, 2-bromoethoxy methyl, 1-bromo-2-benzyloxyethane, 2-bromoethoxymethyl benzene, 2-bromo-ethoxymethyl-benzene | |
| 2-bromoethoxymethylbenzene |
Specifications
| 1462-37-9 | |
| 95°C | |
| MFCD01321307 | |
| benzyl 2-bromoethyl ether, 2-bromoethoxy methyl benzene, 2-benzyloxy-1-bromoethane, benzyloxyethyl bromide, benzene, 2-bromoethoxy methyl, 1-bromo-2-benzyloxyethane, 2-bromoethoxymethyl benzene, 2-bromo-ethoxymethyl-benzene | |
| C1=CC=C(C=C1)COCCBr | |
| 215.09 | |
| 215.09 | |
| Liquid |
| Yellow | |
| C9H11BrO | |
| 5 g | |
| FWOHDAGPWDEWIB-UHFFFAOYSA-N | |
| 2-bromoethoxymethylbenzene | |
| 73833 | |
| ≥97.0% (GC) | |
| Benzyl 2-Bromoethyl Ether |
Safety and Handling
TSCA : No