missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-bromoethyl ether, 97%

Catalog No. AAH5687206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5687206 5 g
AAH5687214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5687206 Supplier Thermo Scientific Chemicals Supplier No. H5687206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1462-37-9 | C9H11BrO | 215.09 g/mol

Alkylation reagent for synthesis of a macrocyclic antifungal antibiotic. Building block used to prepare a copper phthalocyanine discotic liquid crystal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Alkylation reagent for synthesis of a macrocyclic antifungal antibiotic. Building block used to prepare a copper phthalocyanine discotic liquid crystal.

Solubility
Slightly soluble in water

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.

Chemical Identifiers

CAS 1462-37-9
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD01321307
InChI Key FWOHDAGPWDEWIB-UHFFFAOYSA-N
Synonym benzyl 2-bromoethyl ether, 2-bromoethoxy methyl benzene, 2-benzyloxy-1-bromoethane, benzyloxyethyl bromide, benzene, 2-bromoethoxy methyl, 1-bromo-2-benzyloxyethane, 2-bromoethoxymethyl benzene, 2-bromo-ethoxymethyl-benzene
PubChem CID 73833
IUPAC Name 2-bromoethoxymethylbenzene
SMILES C1=CC=C(C=C1)COCCBr

Specifications

Density 1.36
Boiling Point 254°C
Flash Point >113°C (235°F)
Refractive Index 1.541
Linear Formula C6H5CH2OCH2CH2Br
Quantity 5 g
Solubility Information Slightly soluble in water
Formula Weight 215.09
Percent Purity 97%
Chemical Name or Material Benzyl 2-bromoethyl ether
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only