missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2,4-Dihydroxyphenyl Ketone 98.0+%, TCI America™

Catalog No. p-7123246
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B39215G Supplier TCI America Supplier No. B39215G
Only null left

Chemical Identifiers

CAS 3669-41-8
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.25
MDL Number MFCD00043754
InChI Key VFQKAJVKZKHVPD-UHFFFAOYSA-N
Synonym 1-2,4-dihydroxyphenyl-2-phenylethanone, 2',4'-dihydroxy-2-phenylacetophenone, benzyl 2,4-dihydroxyphenyl ketone, 1-2,4-dihydroxyphenyl-2-phenylethan-1-one, 2,4-dihydroxyphenylbenzyl ketone, 2,4-dihydroxydeoxybenzoin, acmc-20aod0, cbmicro_026224, cambridge id 5938997, benzyl2,4-dihydroxyphenylketone
PubChem CID 138005
IUPAC Name 1-(2,4-dihydroxyphenyl)-2-phenylethan-1-one
SMILES OC1=CC=C(C(=O)CC2=CC=CC=C2)C(O)=C1

Specifications

Melting Point 113°C
Boiling Point 225°C
Quantity 5 g
Formula Weight 228.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl 2,4-Dihydroxyphenyl Ketone