missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2,2,2-Trifluoro-N-phenylacetimidate 98.0+%, TCI America™

Catalog No. B32345G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32345G 5 g
B32341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32345G Supplier TCI America Supplier No. B32345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 952057-61-3
Molecular Formula C15H12F3NO
Molecular Weight (g/mol) 279.262
InChI Key KVGZVQNJUZHPJJ-UHFFFAOYSA-N
Synonym 2,2,2-Trifluoro-N-phenylacetimidic Acid Benzyl Ester
PubChem CID 23583338
IUPAC Name benzyl 2,2,2-trifluoro-N-phenylethanimidate
SMILES C1=CC=C(C=C1)COC(=NC2=CC=CC=C2)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 279.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl 2,2,2-Trifluoro-N-phenylacetimidate
TSCA No
Recommended Storage Refrigerator