accessibility menu, dialog, popup

UserName

CAS RN 952057-61-3

IUPAC Name: benzyl 2,2,2-trifluoro-N-phenylethanimidate
Synonyms: benzyl 2,2,2-trifluoro-N-phenylethanimidate
Molecular Weight (g/mol): 279.26
Molecular Formula: C15H12F3NO
InChi Key: KVGZVQNJUZHPJJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(OCC1=CC=CC=C1)=NC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results