missing translation for 'onlineSavingsMsg'
Learn More

Benzoylformic Acid 97.0+%, TCI America™

Catalog No. p-7118867
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B11295G 5 g
B112925G 25 g
2 options

Catalog No. B11295G

Supplier: TCI America B11295G

Only null left

Chemical Identifiers

CAS 611-73-4
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00002575
InChI Key FAQJJMHZNSSFSM-UHFFFAOYSA-N
Synonym benzoylformic acid, phenylglyoxylic acid, oxophenylacetic acid, benzeneglyoxylic acid, phenylgloxylic acid, 2-phenylethanoic acid, oxo phenyl acetic acid, benzoylformate, phenylglyoxalate, phenyloxoacetic acid
PubChem CID 11915
ChEBI CHEBI:18280
IUPAC Name 2-oxo-2-phenylacetic acid
SMILES OC(=O)C(=O)C1=CC=CC=C1

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 138°C
Quantity 5 g
Formula Weight 150.13
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoylformic Acid
RTECSNumber DA1480000
TSCA No