missing translation for 'onlineSavingsMsg'
Learn More

Benzoylformic acid, 97%

Catalog No. p-4509384
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105760050 5 g Glass bottle
AC105761000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105760050 Supplier Thermo Scientific Chemicals Supplier No. 105760050
Only null left

CAS: 611-73-4 | C8H6O3 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-73-4
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.13
MDL Number MFCD00002575
InChI Key FAQJJMHZNSSFSM-UHFFFAOYSA-N
Synonym benzoylformic acid, phenylglyoxylic acid, oxophenylacetic acid, benzeneglyoxylic acid, phenylgloxylic acid, 2-phenylethanoic acid, oxo phenyl acetic acid, benzoylformate, phenylglyoxalate, phenyloxoacetic acid
PubChem CID 11915
ChEBI CHEBI:18280
IUPAC Name 2-oxo-2-phenylacetic acid
SMILES OC(=O)C(=O)C1=CC=CC=C1

Specifications

Melting Point 60°C to 69°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5COCO2H
Quantity 5 g
Beilstein 10,654
Solubility Information Solubility in water: .. Other solubilities: 1g/10mL methanol
Formula Weight 150.13
Percent Purity 97%
Physical Form Adhering Crystals, Crystalline Powder or Chunks
Chemical Name or Material Benzoylformic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 210-278-7

RUO – Research Use Only