missing translation for 'onlineSavingsMsg'
Learn More

Benzoylferrocene 98.0+%, TCI America™

Catalog No. b08865g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08865G 5 g
B088625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08865G Supplier TCI America Supplier No. B08865G
Only null left

Chemical Identifiers

CAS 1272-44-2
Molecular Formula C17H14FeO
Molecular Weight (g/mol) 290.14
MDL Number MFCD00001431
InChI Key JJHHJZCYDPLHIL-UHFFFAOYNA-N
IUPAC Name Benzoylferrocene
SMILES [Fe].c1cccc1.O=C(c1cccc1)C1=CC=CC=C1

Specifications

Melting Point 110°C
Quantity 5 g
Formula Weight 290.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzoylferrocene