accessibility menu, dialog, popup

UserName

FeO

CAS RN 1272-44-2

IUPAC Name: Benzoylferrocene
Synonyms:
Molecular Weight (g/mol): 290.14
Molecular Formula: C17H14FeO
InChi Key:
SMILES: [Fe].c1cccc1.O=C(c1cccc1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results