missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoyl Fluoride 98.0+%, TCI America™
Supplier: TCI America B111525G
Specifications
| Benzoyl Fluoride | |
| -28°C | |
| C7H5FO | |
| 25 g | |
| benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa | |
| FC(=O)C1=CC=CC=C1 | |
| 124.11 | |
| 124.11 | |
| Liquid |
| 455-32-3 | |
| 160°C | |
| MFCD00000364 | |
| 2920 | |
| HPMLGNIUXVXALD-UHFFFAOYSA-N | |
| benzoyl fluoride | |
| 67999 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 455-32-3 | |
| 124.11 | |
| HPMLGNIUXVXALD-UHFFFAOYSA-N | |
| 67999 | |
| FC(=O)C1=CC=CC=C1 |
| C7H5FO | |
| MFCD00000364 | |
| benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa | |
| benzoyl fluoride |
Safety and Handling
TSCA : Yes