missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl Fluoride 98.0+%, TCI America™

Catalog No. p-7118859
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11155G 5 g
B111525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11155G Supplier TCI America Supplier No. B11155G
Only null left

Chemical Identifiers

CAS 455-32-3
Molecular Formula C7H5FO
Molecular Weight (g/mol) 124.11
MDL Number MFCD00000364
InChI Key HPMLGNIUXVXALD-UHFFFAOYSA-N
Synonym benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa
PubChem CID 67999
IUPAC Name benzoyl fluoride
SMILES FC(=O)C1=CC=CC=C1

Specifications

Melting Point -28°C
Boiling Point 160°C
Quantity 5 g
UN Number 2920
Formula Weight 124.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Benzoyl Fluoride