missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoyl-DL-valine 98.0+%, TCI America™
$52.19 - $495.84
Chemical Identifiers
| CAS | 2901-80-6 |
|---|---|
| Molecular Formula | C12H15NO3 |
| Molecular Weight (g/mol) | 221.256 |
| MDL Number | MFCD00038283 |
| InChI Key | MIYQNOPLWKCHED-UHFFFAOYSA-N |
| PubChem CID | 226682 |
| IUPAC Name | 2-benzamido-3-methylbutanoic acid |
| SMILES | CC(C)C(C(=O)O)NC(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 2901-80-6 | |
| 221.256 | |
| MIYQNOPLWKCHED-UHFFFAOYSA-N | |
| 2-benzamido-3-methylbutanoic acid |
| C12H15NO3 | |
| MFCD00038283 | |
| 226682 | |
| CC(C)C(C(=O)O)NC(=O)C1=CC=CC=C1 |
Specifications
| 2901-80-6 | |
| White | |
| MFCD00038283 | |
| MIYQNOPLWKCHED-UHFFFAOYSA-N | |
| 2-benzamido-3-methylbutanoic acid | |
| 226682 | |
| ≥98.0% (T) | |
| Benzoyl-DL-valine |
| 132°C | |
| C12H15NO3 | |
| 1 g | |
| CC(C)C(C(=O)O)NC(=O)C1=CC=CC=C1 | |
| 221.256 | |
| 221.26 | |
| Crystalline Powder |
Safety and Handling
TSCA : No